Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ethyl 7-(diethylamino)-2-oxo-2H-chromene-3-carboxylate | 28705-46-6 | MFCD00227484 | 99.8% | 289.33 g/mol | C16H19NO4 | 250 MG
SDP

Catalog No. 5000209228
Encompass_Preferred

Ethyl 7-(diethylamino)-2-oxo-2H-chromene-3-carboxylate is a chromene-derived research compound reported as a selective inhibitor of human monoamine oxidase B (hMAO-B) with a reported IC50 of 45.52 μM. It is supplied as a high-purity solid for laboratory research applications.

  • Reported IC50 of 45.52 μM against hMAO-B.
  • High purity: 99.8% (as supplied).
  • Physical form: solid, light green to green.
  • Available in multiple pack sizes, including 250 mg.
  • Recommended storage: powder -20°C (up to 3 years) or 4°C (up to 2 years).

Catalog No. 50-002-09228 Supplier Medchemexpress LLC Supplier No. 250MGHYW087433
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ethyl 7-(diethylamino)-2-oxo-2H-chromene-3-carboxylate is a chromene-derived research compound reported as a selective inhibitor of human monoamine oxidase B (hMAO-B) with a reported IC50 of 45.52 μM. It is supplied as a high-purity solid for laboratory research applications.

  • Reported IC50 of 45.52 μM against hMAO-B.
  • High purity: 99.8% (as supplied).
  • Physical form: solid, light green to green.
  • Available in multiple pack sizes, including 250 mg.
  • Recommended storage: powder -20°C (up to 3 years) or 4°C (up to 2 years).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.