Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl Acetate:2-Propanol, 2:1, Reagents

Catalog No. 01335404
Change view
Click to view available options
Quantity:
4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-335-404 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-335-404 Supplier Reagents Holdings Llc Supplier No. CE0015004C
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ethyl Acetate:2-Propanol, 2:1, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

CAS 67-63-0, 141-78-6
Color Colorless
Quantity 4 L
Packaging Glass Bottle
Physical Form Liquid
Specific Gravity 0.8567
Molecular Formula C3H8O
Boiling Point 80°C
InChI Key KFZMGEQAYNKOFK-UHFFFAOYSA-N
SMILES CC(C)O
IUPAC Name propan-2-ol
Molecular Weight (g/mol) 60.10
Grade Reagent
Chemical Name or Material Ethyl Acetate:2-Propanol, 2:1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.