Learn More
U.S. Pharmacopeia Ethyl Acetate, 141-78-6, MFCD00009171, 1.2 mL/ampule; 3 ampules
Catalog No. NC1409120
Encompass
Molecular formula C4H8O2, Molecular weight 88.11, Melting point/freezing point -117.4 °F (-83 °C), Initial boiling point 170.6 °F (77 °C), Synonyms: Acetic ether, Ethyl ethanoate
Catalog No. NC1409120 Supplier U.S. Pharmacopeia Supplier No. 1265402
Molecular formula C4H8O2, Molecular weight 88.11, Melting point/freezing point -117.4 °F (-83 °C), Initial boiling point 170.6 °F (77 °C), Synonyms: Acetic ether, Ethyl ethanoate