Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl acetate, 99.8%, for biochemistry, AcroSeal™, ACROS Organics™

Click to view available options
Quantity:
1L
2.5L
Packaging:
AcroSeal™ Glass bottle
141-78-6
C4H8O2
88.106
XEKOWRVHYACXOJ-UHFFFAOYSA-N
ethyl ethanoate,acetoxyethane,acetic acid ethyl ester,acetic ether,ethyl acetic ester,vinegar naphtha,acetic acid, ethyl ester,ethylacetate,acetidin,acetic ester
8857
CHEBI:27750
ethyl acetate
CCOC(=O)C
141-78-6
C4H8O2
88.106
XEKOWRVHYACXOJ-UHFFFAOYSA-N
ethyl ethanoate,acetoxyethane,acetic acid ethyl ester,acetic ether,ethyl acetic ester,vinegar naphtha,acetic acid, ethyl ester,ethylacetate,acetidin,acetic ester
8857
CHEBI:27750
ethyl acetate
CCOC(=O)C
Absorbance 0.03 max. at 280nm,0.05 max. at 265nm,0.1 max. at 263nm,0.3 max. at 260nm,1 max. at 255nm
Assay Percent Range 99.8%
Linear Formula CH3CO2C2H5
Solubility Information Solubility in water: 80g/L (20°C). Other solubilities: soluble in benzene, acetone, chloroform, alcohol,, ether
Formula Weight 88.11
Percent Purity 99.8%
Grade Biochemical
Density 0.9020g/mL
Packaging AcroSeal™ Glass bottle
Refractive Index 1.3710 to 1.3740
Solubility Solubility in water: 80g/L (20°C). Other solubilities: soluble in benzene, acetone, chloroform, alcohol,, ether
Specific Gravity 0.902
Flash Point −4°C
Viscosity 0.45 mPa.s (20°C)
Boiling Point 75.0°C to 78.0°C
Melting Point -83.5°C
Quantity 1L
Chemical Name or Material Ethyl acetate
Show More Show Less