Learn More
Sigma Aldrich Fine Chemicals Biosciences Ethyl acetate 99 fCC fG
Catalog No. 502899677
Encompass
Ethyl acetate (EtOAc) is an effective alternate solvent for diethyl ether employed for the concentration of eggs larvae and cysts in fecal specimens during the formalin-ether sedimentation technique. Acetaldehyde is reaction intermediate formed during the oxidative combustion of ethanol and ethyl acetate in the presence of copper oxide embedded on Ce-doped titania surface.
Catalog No. 50-289-9677 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. W24140710KGK
Ethyl acetate (EtOAc) is an effective alternate solvent for diethyl ether employed for the concentration of eggs larvae and cysts in fecal specimens during the formalin-ether sedimentation technique. Acetaldehyde is reaction intermediate formed during the oxidative combustion of ethanol and ethyl acetate in the presence of copper oxide embedded on Ce-doped titania surface.