Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl Acetate ACS, MilliporeSigma™

Catalog No. MEX02409 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 L
4 L
Packaging:
Poly Bottle
Poly-coated Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MEX02409 1 L Poly Bottle
MEX0240P4 4 L Poly-coated Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MEX02409 Supplier MilliporeSigma Supplier No. EX02409
Add to Cart
Add to Cart

Chemical Identifiers

CAS 141-78-6
Molecular Formula C4H8O2
Molecular Weight (g/mol) 88.106
InChI Key XEKOWRVHYACXOJ-UHFFFAOYSA-N
Synonym ethyl ethanoate, acetoxyethane, acetic acid ethyl ester, acetic ether, ethyl acetic ester, vinegar naphtha, acetic acid, ethyl ester, ethylacetate, acetidin, acetic ester
PubChem CID 8857
ChEBI CHEBI:27750
IUPAC Name ethyl acetate
SMILES CCOC(=O)C

Specifications

Melting Point -84°C
Color Colorless
Quantity 1 L
Flash Point -4°C (-25°F)
Ignition Point 425.85°C (798.5°F)
Odor Charactristic
Packaging Poly Bottle
Physical Form Liquid
Linear Formula CH3COOCH2CH3
UN Number UN 1173
Boiling Point 77.2°C
Solubility Information Partially soluble in water
Formula Weight 88.11
Percent Purity ≥99.5%
Grade ACS
Chemical Name or Material Ethyl Acetate
Show More Show Less
DOTInformation DOT Class: 3, Packaging Group: II
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.