Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl Acetate, ACS Reagent, for HPLC, Spectrophotometry, meets analytical spec. of USP, ≥99.5%, Solstice

Catalog No. 6000589 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
4 x 4 L
Packaging:
Glass Bottle
Poly-coated Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-005-89 4 x 4 L Glass Bottle
60-005-90 4 x 4 L Poly-coated Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-005-89 Supplier Solstice Supplier No. AH1004
Add to Cart
Add to Cart

Chemical Identifiers

CAS 141-78-6
Molecular Formula C4H8O2
Molecular Weight (g/mol) 88.106
MDL Number MFCD00009171
InChI Key XEKOWRVHYACXOJ-UHFFFAOYSA-N
Synonym ethyl ethanoate, acetoxyethane, acetic acid ethyl ester, acetic ether, ethyl acetic ester, vinegar naphtha, acetic acid, ethyl ester, ethylacetate, acetidin, acetic ester
PubChem CID 8857
ChEBI CHEBI:27750
IUPAC Name ethyl acetate
SMILES CCOC(=O)C

Specifications

CAS Max % 100.0000%
Density 0.8945 g/cm3
Quantity 4 x 4 L
Flash Point −4°C
Packaging Glass Bottle
Water 0.03% max.
UN Number UN1173
Beilstein 506104
Boiling Point 77°C
Solubility Information 870% in Water at 20°C
Formula Weight 88.11
Percent Purity >99.5%
Grade HPLC
Chemical Name or Material Ethyl Acetate, ACS Reagent, for HPLC, Spectrophotometry, meets analytical specification of USP, ≥99.5%
Show More Show Less
ShelfLife 547 days from date of manufacture
DOTInformation Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Ethyl acetate
EINECSNumber 205-500-4
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.