Learn More
Ethyl Acetate, ACS Reagent, Reagents
Specifications
Specifications
| Melting Point | -83.6°C |
| Color | Colorless |
| Quantity | 4 L |
| Packaging | Glass Bottle |
| Physical Form | Liquid |
| Specific Gravity | 0.8946 |
| Boiling Point | 77°C |
| Grade | ACS |
| Chemical Name or Material | Ethyl Acetate, ACS Reagent |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.