Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl Acetate, ACS Reagent, Reagents

Catalog No. 01337991
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-337-991 20 L
01-337-992 1 L
01-337-990 4 L
01-337-989 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 01-337-991 Supplier Reagents Holdings Llc Supplier No. C119950020E6
Only null left
Add to Cart
Add to Cart

Ethyl Acetate, ACS Reagent, ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 141-78-6
Molecular Formula C4H8O2
Molecular Weight (g/mol) 88.11
InChI Key XEKOWRVHYACXOJ-UHFFFAOYSA-N
Synonym Ethyl Ethanoate, Acetic Acid Ethyl Ester, Acetic Ether
IUPAC Name ethyl acetate
SMILES CCOC(C)=O

Specifications

Melting Point -83.6°C
Color Colorless
Quantity 20 L
Packaging Steel Pail
Physical Form Liquid
Specific Gravity 0.8946
Boiling Point 77°C
Grade ACS
Chemical Name or Material Ethyl Acetate, ACS Reagent
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.