Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl Acetate Biosynthesis, 99.9%, MilliporeSigma™

Catalog No. 50947983 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
500 mL
4 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-947-983 500 mL
50-947-989 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-947-983 Supplier MilliporeSigma Supplier No. EX02425
Add to Cart
Add to Cart

Chemical Identifiers

CAS 141-78-6
Molecular Formula C4H8O2
Molecular Weight (g/mol) 88.11
InChI Key XEKOWRVHYACXOJ-UHFFFAOYSA-N
IUPAC Name ethyl acetate
SMILES CCOC(C)=O

Specifications

Melting Point −83°C
Quantity 500 mL
Flash Point −4°C
Packaging Glass Bottle (Pre-packed)
Residue after Evaporation 1 ppm max
Assay Percent Range 99.9% min
Vapor Pressure 97 hPa (20°C)
Boiling Point 77°C (1013 hPa)
Solubility Information 85.3 g/L
Recommended Storage Store at room temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.