Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl Acetate, Biotech Grade, ≥99.8%,Solstice

Catalog No. 60046994 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
2 L
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-046-994 2 L Glass Bottle
60-046-993 1 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-046-994 Supplier Solstice Supplier No. 4945182L
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Biotech. grade,≥99.8%

  • Solstice developed the first high-purity solvents more than 50 years ago
  • As new applications and techniques emerge, Solstice continues to develop products to meet the most demanding requirements for laboratories and pharmaceutical production
  • Our packaging solutions, ranging from lab scale through production, provide our customers with added value, increased efficiency, and enhanced safety.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 141-78-6
Molecular Formula C4H8O2
Molecular Weight (g/mol) 88.106
MDL Number MFCD00009171
InChI Key XEKOWRVHYACXOJ-UHFFFAOYSA-N
Synonym ethyl ethanoate, acetoxyethane, acetic acid ethyl ester, acetic ether, ethyl acetic ester, vinegar naphtha, acetic acid, ethyl ester, ethylacetate, acetidin, acetic ester
PubChem CID 8857
ChEBI CHEBI:27750
IUPAC Name ethyl acetate
SMILES CCOC(=O)C

Specifications

Melting Point -84°C
Color Colorless
Density 0.902g/mL at 25°C
Quantity 2 L
Explosion Limit 2.2 to 11.5% (38°F)
Flash Point −3°C (26.6°F)
Ignition Point 801°F
Odor Characteristic fruity
Packaging Glass Bottle
Physical Form Liquid
Linear Formula CH3COOC2H5
Vapor Pressure 73mmHg (20°C)
UN Number UN1173
Beilstein 506104
Boiling Point 77°C
Formula Weight 88.11g/mol
Percent Purity ≥99.8%
Chemical Name or Material Ethyl acetate
Show More Show Less
Hazard Statement P210-P261-P305 + P351 + P338
DOTInformation class 3 - PG 2 - Ethyl acetate
RTECSNumber 205-500-4
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.