Learn More
Sigma Aldrich Fine Chemicals Biosciences Ethyl acetate/Ethanol 31
Catalog No. 502905169
Encompass
We are committed to bringing you Greener Alternative Products which adhere to one or more of The 12 Principles of Greener Chemistry. This SOLUTIONon of Ethyl acetate/Ethanol is a greener alternative to dichloromethane for chromatographic purification and thus has been enhanced for Safer Solvents and Auxiliaries. Click here for more information.
Catalog No. 50-290-5169 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 74558820L
We are committed to bringing you Greener Alternative Products which adhere to one or more of The 12 Principles of Greener Chemistry. This SOLUTIONon of Ethyl acetate/Ethanol is a greener alternative to dichloromethane for chromatographic purification and thus has been enhanced for Safer Solvents and Auxiliaries. Click here for more information.