Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl acetate, For HPLC, ≥99.8%, MilliporeSigma™ Supelco™

Catalog No. 111005332 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
50 L
Packaging:
Composite Drum
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
11-100-5332 50 L Composite Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-100-5332 Supplier MilliporeSigma Supelco Supplier No. 43916950LP23A
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ethyl acetate (EA), a carboxylate ester, is bio-friendly organic solvent with a wide range of industrial applications. Its synthesis by reactive distillation and by acceptorless dehydrogenative dimerization of ethanol has been explored. Its ability as an acyl acceptor in the immobilized lipase-mediated preparation of biodiesel from crude vegetable oils has been examined. The complete degradation of ethyl acetate to CO2 using manganese octahedral molecular sieve (OMS-2) has been investigated. EA is an effective alternate solvent of diethyl ether, employed for the concentration of eggs, larvae, and cysts in fecal specimens during the Formalin-ether sedimentation technique. Acetaldehyde is reaction intermediate formed during the oxidative combustion of ethanol and ethyl acetate in the presence of copper oxide embedded on Ce-doped titania surface.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 141-78-6
Molecular Formula C4H8O2
Molecular Weight (g/mol) 88.11
MDL Number MFCD00009171
InChI Key XEKOWRVHYACXOJ-UHFFFAOYSA-N
Synonym EtOAc
IUPAC Name ethyl acetate
SMILES CCOC(C)=O

Specifications

Melting Point -84°C
Density 0.902 g/mL (at 25°C)
Quantity 50 L
Packaging Composite Drum
Refractive Index n20/D 1.3720
Vapor Pressure 73 mmHg (20°C)
Linear Formula CH3COOC2H5
Boiling Point 76.5°C to 77.5°C
Formula Weight 88.11
Percent Purity ≥99.8%
Grade HPLC
Show More Show Less
RTECSNumber AH5425000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.