Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl Acetate, Solstice

Catalog No. 60090198 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
56 L
Packaging:
Returnable Stainless Steel Drum
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-090-198 56 L Returnable Stainless Steel Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60-090-198 Supplier Solstice Supplier No. CS100ZA56
Add to Cart
Add to Cart

ACS Reagent, for organic synthesis, prep-LC, and general laboratory use, >99.5%

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 141-78-6
PubChem CID 8857
ChEBI CHEBI:27750

Specifications

Quantity 56 L
Flash Point -4 °C
Packaging Returnable Stainless Steel Drum
Physical Form Liquid, Clear
Grade ACS
Recommended Storage Room Temperature
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.