Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl Acetate, HPLC Grade, MilliporeSigma™

Catalog No. MEX0245P1 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
4 L
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MEX0245P1 4 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MEX0245P1 Supplier MilliporeSigma Supplier No. US1EX0245P/1
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 141-78-6
Molecular Formula C4H8O2
Molecular Weight (g/mol) 88.106
InChI Key XEKOWRVHYACXOJ-UHFFFAOYSA-N
Synonym ethyl ethanoate, acetoxyethane, acetic acid ethyl ester, acetic ether, ethyl acetic ester, vinegar naphtha, acetic acid, ethyl ester, ethylacetate, acetidin, acetic ester
PubChem CID 8857
ChEBI CHEBI:27750
IUPAC Name ethyl acetate
SMILES CCOC(=O)C

Specifications

Melting Point -83°C
Quantity 4 L
Flash Point -4°C
Packaging Glass Bottle
Physical Form Clear liquid, free from particulates
Residue after Evaporation 5ppm
Titratable Acid 0.0009meq/g
Vapor Pressure 97 hPa (20°C)
Water 0.05%
Boiling Point 77°C (1013 hPa)
Solubility Information 85.3 g/L
Formula Weight 88.11
Percent Purity 99.8%
Chemical Name or Material Ethyl Acetate
Show More Show Less
EINECSNumber 205-500-4
RTECSNumber AH5425000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.