Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl Acetate, Innovating Science™
SCI_ED

Catalog No. S031127 Shop All Aldon Products
Change view
Click to view available options
Quantity:
30 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S031127 500 mL
S031142 30 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S031127 Supplier Aldon Corporation Supplier No. IS14035
Add to Cart
Add to Cart

Chemical Identifiers

Molecular Weight (g/mol) 88.11 g/mol

Specifications

Quantity 500 mL
Packaging Steel Conetop Can
Physical Form Liquid
Boiling Point 77°C
Percent Purity 99.5%
Chemical Name or Material Ethyl Acetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.