Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl Acetate, Purified, Reagents

Catalog No. 01337262
Change view
Click to view available options
Quantity:
500 mL
4 L
20 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-337-262 500 mL
01-337-264 20 L
01-337-263 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01-337-262 Supplier Reagents Holdings Llc Supplier No. C2199500500C
Add to Cart
Add to Cart

Ethyl Acetate, Purified, Chemically Pure, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 141-78-6
Molecular Formula C4H8O2
Molecular Weight (g/mol) 88.11
InChI Key XEKOWRVHYACXOJ-UHFFFAOYSA-N
Synonym Ethyl Ethanoate, Acetic Acid Ethyl Ester, Acetic Ether
IUPAC Name ethyl acetate
SMILES CCOC(C)=O

Specifications

Melting Point -83.6°C
Color Colorless
Quantity 500 mL
Packaging Glass Bottle
Physical Form Liquid
Specific Gravity 0.8946
Boiling Point 77°C
Grade Pure
Chemical Name or Material Ethyl Acetate, Purified
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.