Learn More
Ethyl Acetate, Purified, Reagents
Specifications
Specifications
| Melting Point | -83.6°C |
| Color | Colorless |
| Quantity | 4 L |
| Packaging | Glass Bottle |
| Physical Form | Liquid |
| Specific Gravity | 0.8946 |
| Boiling Point | 77°C |
| Grade | Pure |
| Chemical Name or Material | Ethyl Acetate, Purified |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.