Learn More
Sigma Aldrich Fine Chemicals Biosciences Ethyl acetate suitable for
Catalog No. 502897307
Encompass
Ethyl acetate is an effective alternate solvent of diethyl ether.
Catalog No. 50-289-7307 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 348582L
Ethyl acetate is an effective alternate solvent of diethyl ether.