Learn More
Sigma Aldrich Fine Chemicals Biosciences Ethyl acetate suitable for
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 4391694X4L
Encompass
Ethyl acetate (EA) a carboxylate ester is bio-friendly organic solvent with a wide range of industrial applications. Its synthesis by reactive distillation and by acceptorless dehydrogenative dimerization of ethanol has been explored. Its ability as an acyl acceptor in the immobilized lipase-mediated preparation of biodiesel from crude vegetable oils has been examined. The complete degradation of ethyl acetate to CO2 using manganese octahedral molecular sieve (OMS-2) has been investigated. EA is an effective alternate solvent of diethyl ether employed for the concentration of eggs larvae and cysts in fecal specimens during the formalin-ether sedimentation technique. Acetaldehyde is reaction intermediate formed during the oxidative combustion of ethanol and ethyl acetate in the presence of copper oxide embedded on Ce-doped titania surface.
Catalog No. 50-290-3384