Learn More
Sigma Aldrich Fine Chemicals Biosciences Ethyl acetate suitable for
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 348584X2L
Encompass
Ethyl acetate is an effective alternate solvent of diethyl ether.
Catalog No. 50-289-7540