Learn More
Sigma Aldrich Fine Chemicals Biosciences Ethyl acetate suitable for1L
Catalog No. 502898318
Encompass
Ethyl acetate is an effective alternate solvent of diethyl ether.
Catalog No. 50-289-8318 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 348581L
Ethyl acetate is an effective alternate solvent of diethyl ether.