Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl Acetate, Ultra Pure Grade, MP Biomedicals™

Catalog No. 0230008416 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
4 x 4 L
6 x 1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
02-300-08416 4 x 4 L
02-300-08406 6 x 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 02-300-08416 Supplier MP Biomedicals, Inc Supplier No. 0230008416
Add to Cart
Add to Cart

Ethyl Acetate, Ultra Pure Grade is a solvent and diluent.

  • Ethyl Acetate is suitable for HPLC, spectrophotometry, environmental testing.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 141-78-6
Molecular Formula C4H8O2
Molecular Weight (g/mol) 88.11
InChI Key XEKOWRVHYACXOJ-UHFFFAOYSA-N
IUPAC Name ethyl acetate
SMILES CCOC(C)=O

Specifications

Melting Point -118.5°F
Quantity 4 x 4 L
Boiling Point 171°F (760 mmHg)
Percent Purity ≥99.9%
Grade Ultra Pure

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.