Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl Acetate, ULTRA RESI-ANALYZED™, J.T. Baker™

Catalog No. 02002828 Shop All Avantor Products
Click to view available options
Quantity:
1 L
4 L
Packaging:
Narrow Mouth Amber Glass Bottle

Catalog No. 02-002-828

Supplier: Avantor 926002

Only null left
Add to Cart
Add to Cart

High purity solvents and salts developed for organic residue extraction/concentration procedures

    For use in Gas Chromatography (GC), Organic Residue Analysis

Chemical Identifiers

CAS 141-78-6
Molecular Formula C4H8O2
Molecular Weight (g/mol) 88.106
InChI Key XEKOWRVHYACXOJ-UHFFFAOYSA-N
Synonym ethyl ethanoate, acetoxyethane, acetic acid ethyl ester, acetic ether, ethyl acetic ester, vinegar naphtha, acetic acid, ethyl ester, ethylacetate, acetidin, acetic ester
PubChem CID 8857
ChEBI CHEBI:27750
IUPAC Name ethyl acetate
SMILES CCOC(=O)C

Specifications

Quantity 1 L
Flash Point −4.00°C (− 39.2°F)
Packaging Narrow Mouth Amber Glass Bottle
Specific Gravity 1L = 0.90kg
Formula Weight 88.11
Grade ULTRA RESI-ANALYZED
Chemical Name or Material Ethyl Acetate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.