Learn More
Medchemexpress LLC Ethyl acetoacetate-13C | 61973-42-0 | 98.6% | 131.13 | 100 MG
SDP
Catalog No. 5000364745
Encompass_Preferred
Ethyl acetoacetate-13C is a 13C labeled ester extensively used as an intermediate in the synthesis of various compounds. It is also identified as an inhibitor of bacterial biofilm, and can be utilized as a tracer and an internal standard for quantitative analysis via methods such as NMR, GC-MS, or LC-MS.
  • Used as a tracer
  • Serves as an internal standard for quantitative analysis
  • Incorporates carbon-13, a stable heavy isotope, for quantitation during drug development
Catalog No. 50-003-64745 Supplier Medchemexpress LLC Supplier No. 100MGHYY1093S3
Ethyl acetoacetate-13C is a 13C labeled ester extensively used as an intermediate in the synthesis of various compounds. It is also identified as an inhibitor of bacterial biofilm, and can be utilized as a tracer and an internal standard for quantitative analysis via methods such as NMR, GC-MS, or LC-MS.
  • Used as a tracer
  • Serves as an internal standard for quantitative analysis
  • Incorporates carbon-13, a stable heavy isotope, for quantitation during drug development