Learn More
Medchemexpress LLC Ethyl acetoacetate-13C | 61973-42-0 | 98.6% | 131.13 | 500 MG
SDP
Catalog No. 5000365885
Encompass_Preferred
Ethyl acetoacetate-13C is the 13C labeled Ethyl acetoacetate. Ethyl acetoacetate (Ethyl acetylacetate) is an ester widely used as an intermediate in the synthesis of many varieties of compounds.
  • Inhibitor of bacterial biofilm.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-003-65885 Supplier Medchemexpress LLC Supplier No. 500MGHYY1093S3
Ethyl acetoacetate-13C is the 13C labeled Ethyl acetoacetate. Ethyl acetoacetate (Ethyl acetylacetate) is an ester widely used as an intermediate in the synthesis of many varieties of compounds.
  • Inhibitor of bacterial biofilm.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.