Learn More
Medchemexpress LLC Ethyl acetoacetate | 141-97-9 | MFCD00009199 | 99.5% | 130.14 g/mol | C6H10O3 | 100g
SDP
Catalog No. 5000267090
Encompass_Preferred
Ethyl acetoacetate (Ethyl acetylacetate) is an ester widely used as an intermediate in the synthesis of many varieties of compounds[1][2][3] Ethyl acetoacetate is an inhibitor of bacterial biofilm[4]
Catalog No. 50-002-67090 Supplier Medchemexpress LLC Supplier No. HYY1093100G
Ethyl acetoacetate (Ethyl acetylacetate) is an ester widely used as an intermediate in the synthesis of many varieties of compounds[1][2][3] Ethyl acetoacetate is an inhibitor of bacterial biofilm[4]