Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl acetoacetate, ≥99% (GC), MilliporeSigma™ Supelco™

Catalog No. 111019824 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-9824 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-9824 Supplier MilliporeSigma Supelco Supplier No. 639541ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ethyl acetoacetate is derived from ethanol.

Featured Industries: Cleaning Products, Cosmetics, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

CAS 141-97-9
Ignition Point 580°F
Vapor Pressure 1 mmHg (28.5°C)
Vapor Density 4.48 (vs. air)
Density 1.029 g/mL (at 20°C)
Flash Point 73.5°C - closed cup
Physical Form Neat
Linear Formula CH3 COCH2 COOC2H5
MDL Number MFCD00009199
Melting Point -43°C
Boiling Point 181°C
Synonym Acetoacetic ester
Solubility Soluble in water
Type Analytical Standard
Quantity 1 mL
Formula Weight 130.14
Percent Purity ≥99% (GC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.