Learn More
Medchemexpress LLC Ethyl acetoacetate-d3 | 1565868-21-4 | MFCD32204475 | 98.0% | 133.16 | C6H7D3O3 | 25mg
SDP
Catalog No. 5000267126
Encompass_Preferred
Ethyl acetoacetate-d3 is the deuterium labeled Ethyl acetoacetate Ethyl acetoacetate (Ethyl acetylacetate) is an ester widely used as an intermediate in the synthesis of many varieties of compounds[1][2][3] Ethyl acetoacetate is an inhibitor of bacterial biofilm[4]
Catalog No. 50-002-67126 Supplier Medchemexpress LLC Supplier No. HYY1093S425MG
Ethyl acetoacetate-d3 is the deuterium labeled Ethyl acetoacetate Ethyl acetoacetate (Ethyl acetylacetate) is an ester widely used as an intermediate in the synthesis of many varieties of compounds[1][2][3] Ethyl acetoacetate is an inhibitor of bacterial biofilm[4]