Learn More
Medchemexpress LLC Ethyl acetoacetate-d5 | 55514-60-8 | 99.2% | 135.17 | C6H5D5O3 | 5mg
SDP
Catalog No. 5000267473
Encompass_Preferred
Ethyl acetoacetate-d5 is the deuterium labeled Ethyl acetoacetate[1] Ethyl acetoacetate (Ethyl acetylacetate) is an ester widely used as an intermediate in the synthesis of many varieties of compounds[2][3][4] Ethyl acetoacetate is an inhibitor of bacterial biofilm[5]
Catalog No. 50-002-67473 Supplier Medchemexpress LLC Supplier No. HYY1093S25MG
Ethyl acetoacetate-d5 is the deuterium labeled Ethyl acetoacetate[1] Ethyl acetoacetate (Ethyl acetylacetate) is an ester widely used as an intermediate in the synthesis of many varieties of compounds[2][3][4] Ethyl acetoacetate is an inhibitor of bacterial biofilm[5]