Learn More
Medchemexpress LLC Ethyl acetoacetate-d5 | 55514-60-8 | 99.2% | 135.17 g/mol | C6H5D5O3 | 10mg
SDP
Catalog No. 5000267409
Encompass_Preferred
Ethyl acetoacetate-d5 is the deuterium labeled Ethyl acetoacetate[1] Ethyl acetoacetate (Ethyl acetylacetate) is an ester widely used as an intermediate in the synthesis of many varieties of compounds[2][3][4] Ethyl acetoacetate is an inhibitor of bacterial biofilm[5]
Catalog No. 50-002-67409 Supplier Medchemexpress LLC Supplier No. HYY1093S210MG
Add to Cart
Add to Cart
Ethyl acetoacetate-d5 is the deuterium labeled Ethyl acetoacetate[1] Ethyl acetoacetate (Ethyl acetylacetate) is an ester widely used as an intermediate in the synthesis of many varieties of compounds[2][3][4] Ethyl acetoacetate is an inhibitor of bacterial biofilm[5]