Learn More
Sigma Aldrich Fine Chemicals Biosciences Ethyl acetoacetate natural5KG
Catalog No. 502907085
Encompass
Ethyl acetoacetate is ethyl ester of acetoacetic acid. Ethyl acetoacetate exhibits keto-enol tautomerism. Ethyl acetoacetate reacts with aromatic amines in the presence of commercially available zeolite HSZ-360 to form symmetric diphenylureas.
Catalog No. 50-290-7085 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. W2415125KGK
Ethyl acetoacetate is ethyl ester of acetoacetic acid. Ethyl acetoacetate exhibits keto-enol tautomerism. Ethyl acetoacetate reacts with aromatic amines in the presence of commercially available zeolite HSZ-360 to form symmetric diphenylureas.