Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Ethyl acetohydroxamate

Catalog No. 20892210G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
20-892-210G 10 g
20-892-250G 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 20-892-210G Supplier Sigma Aldrich Supplier No. 20892210G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Ethyl acetohydroxamate

Specifications

CAS 10576-12-2
Melting Point 23°C to 25°C (lit.)
Boiling Point 55°C to 58°C (6 mmHg)
Molecular Formula C4H9NO2
Refractive Index n20/D 1.434 (literature)
Linear Formula CH3C(=NOH)OC2H5
MDL Number MFCD00002114
Quantity 10 g
Synonym Ethyl N-hydroxyacetimidate; Ethyl acetohydroximate
Molecular Weight (g/mol) 103.12
Percent Purity 97%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.