Learn More
Medchemexpress LLC Ethyl acetylglycinate | 1906-82-7 | MFCD00009173 | 98.2% | 145.16 g/mol | C6H11NO3 | 100 G
SDP
Catalog No. 5000300054
Encompass_Preferred
Ethyl acetylglycinate is the ethyl ester of N-acetylglycine, an amino acid derivative used as a building block and reagent in synthetic chemistry and biochemical research. It is supplied as a solid with high purity and is suitable for use as an intermediate or for derivatization studies. Molecular formula C6H11NO3; molecular weight 145.16 g/mol.
  • High purity suitable for research applications.
  • Solid form simplifies handling and storage.
  • Useful as a synthetic building block and intermediate.
  • Stable under recommended storage conditions.
  • Available in multiple pack sizes for scalability.
Catalog No. 50-003-00054 Supplier Medchemexpress LLC Supplier No. HYW015651100G
Ethyl acetylglycinate is the ethyl ester of N-acetylglycine, an amino acid derivative used as a building block and reagent in synthetic chemistry and biochemical research. It is supplied as a solid with high purity and is suitable for use as an intermediate or for derivatization studies. Molecular formula C6H11NO3; molecular weight 145.16 g/mol.
  • High purity suitable for research applications.
  • Solid form simplifies handling and storage.
  • Useful as a synthetic building block and intermediate.
  • Stable under recommended storage conditions.
  • Available in multiple pack sizes for scalability.