Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl Acetylsalicylate 98.0+%, TCI America™
SDP

Catalog No. A15925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A15925G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A15925G Supplier TCI America Supplier No. A15925G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 529-68-0
Molecular Formula C11H12O4
Molecular Weight (g/mol) 208.213
MDL Number MFCD00026824
InChI Key UYDSGXAKLVZWIJ-UHFFFAOYSA-N
Synonym 2-Acetoxybenzoic Acid Ethyl Ester, Ethyl 2-Acetoxybenzoate, Acetylsalicylic Acid Ethyl Ester
PubChem CID 10728
IUPAC Name ethyl 2-acetyloxybenzoate
SMILES CCOC(=O)C1=CC=CC=C1OC(=O)C

Specifications

Color Yellow
Quantity 5 g
Formula Weight 208.21
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl Acetylsalicylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.