Learn More
eMolecules​ 140-88-5 | Ethyl Acrylate stabilized with 10 - 20 ppm MEHQ | Oakwood Chemicals | MFCD00009188 | 100.117 | C5H8O2 | 98.000 | CCOC(=O)C=C | 100ml | 480145023
Catalog No. 502697397 Shop All eMolecules Inc. Products
Encompass_Preferred
Ethyl Acrylate stabilized with 10 - 20 ppm MEHQ | Oakwood Chemicals | 140-88-5 | MFCD00009188 | 100.117 | C5H8O2 | 98.000 | CCOC(=O)C=C | 100ml | 480145023
Catalog No. 50-269-7397 Supplier eMolecules​ Supplier No. 480145023
Ethyl Acrylate stabilized with 10 - 20 ppm MEHQ | Oakwood Chemicals | 140-88-5 | MFCD00009188 | 100.117 | C5H8O2 | 98.000 | CCOC(=O)C=C | 100ml | 480145023