Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl Alcohol 140 Proof with Deionized Water, Greenfield

Catalog No. 502946088
Encompass
Change view
Click to view available options
Poly Pail
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
50-294-6088 Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-294-6088 Supplier Thermo Scientific Production Chemicals and Services Supplier No. P111000140PL05
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Chemical Identifiers

CAS 64-17-5 (Ethanol) , 7732-18-5 (Water)
Molecular Weight (g/mol) 46.07 (Ethanol), 18.02 (Water)

Specifications

Quantity 5 Gallon
Packaging Poly Pail
Boiling Point > 172.4 - < 179.6 °F (> 78 - < 82 °C)
Percent Purity 70% (140 Proof)
Chemical Name or Material Ethyl Alcohol 70% (140 Proof)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.