Learn More
Ethyl Alcohol, 190 Proof, Reagent, ACS, 95%, Spectrum™ Chemical
Shop All Spectrum Chemical Manufacturing Corporation Products

Specifications
Specifications
| Color | Colorless |
| CAS Max % | 0.95, 0.05 |
| Appearance | Clear |
| Quantity | 1 L |
| Flash Point | 17°C (63°F) |
| Packaging | Glass Bottle |
| Physical Form | Liquid |
| Specific Gravity | 0.8 at 20°C |
| Concentration | 95% |
| Boiling Point | 79°C |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.