Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl Alcohol 191 Proof USP EP Harmonized, Greenfield

Catalog No. 502946010
Encompass
Change view
Click to view available options
Metal Drum
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
50-294-6010 Metal Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-294-6010 Supplier Thermo Scientific Production Chemicals and Services Supplier No. P1110000EP191DM55M
Add to Cart
Add to Cart

Chemical Identifiers

CAS 64-17-5 (Ethanol)
Molecular Weight (g/mol) 46.07 (Ethanol)

Specifications

Quantity 55 Gallon
Packaging Metal Drum
Boiling Point > 172.4 - < 176 °F (> 78 - < 80 °C)
Percent Purity 95.5% (191 Proof)
Grade USP
Chemical Name or Material Ethyl Alcohol 95.5% (191 Proof)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.