Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl Alcohol 200 Proof ACS USP, Greenfield

Catalog No. 502946014
Encompass
Change view
Click to view available options
Packaging:
Metal Pail
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packaging
50-294-6014 Metal Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-294-6014 Supplier Thermo Scientific Production Chemicals and Services Supplier No. P111000200PL05M
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Chemical Identifiers

CAS 64-17-5
Molecular Weight (g/mol) 46.07

Specifications

Quantity 5 Gallon
Packaging Metal Pail
Boiling Point 172.13 °F (77.85 °C) (101.33 kPa)
Percent Purity 100% (200 Proof)
Grade ACS/NF/USP
Chemical Name or Material Ethyl Alcohol 100% (200 Proof)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.