Learn More
Electron Microscopy Sciences Ethyl Alcohol, Anhydrous, 200 Proof, 64-17-5, 4x450ml, Taxes Paid
SDP
Catalog No. 50980461
Encompass_Preferred
Molecular Formula C2H6O, Molecular Weight 46.068 , Ethanol is avolatile,flammable, colorless liquid with a slight characteristic odor. It is apsychoactive substance. Ethanol is naturally produced by the fermentation of sugars by yeasts or via petrochemical processes. It also has medical applications as an antiseptic and disinfectant. The compound is widely used as a chemical solvent, either for scientific chemical testing or in synthesis of other organic compounds, and is a vital substance used across many different kinds of manufacturing industries. Ethanol is also used as a clean-burning fuel source.
Catalog No. 50-980-461 Supplier Electron Microscopy Sciences Supplier No. 15056
Molecular Formula C2H6O, Molecular Weight 46.068 , Ethanol is avolatile,flammable, colorless liquid with a slight characteristic odor. It is apsychoactive substance. Ethanol is naturally produced by the fermentation of sugars by yeasts or via petrochemical processes. It also has medical applications as an antiseptic and disinfectant. The compound is widely used as a chemical solvent, either for scientific chemical testing or in synthesis of other organic compounds, and is a vital substance used across many different kinds of manufacturing industries. Ethanol is also used as a clean-burning fuel source.