Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl Alcohol 31 Proof, Greenfield

Catalog No. 502946002
Encompass
Change view
Click to view available options
Glass Case
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
50-294-6002 Glass Case
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-294-6002 Supplier Thermo Scientific Production Chemicals and Services Supplier No. P111WFI031CS4L
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Chemical Identifiers

CAS 64-17-5

Specifications

Packaging Case Glass 4x4 L
Boiling Point > 188.6 - < 194 °F (> 87 - < 90 °C)
Percent Purity 15.5% (31 Proof)
Chemical Name or Material Ethyl Alcohol 15.5% (31 Proof)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.