Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl Alcohol 36 Proof With WFI USP, Greenfield

Catalog No. 502946037
Encompass
Change view
Click to view available options
Poly Drum
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
50-294-6037 Poly Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-294-6037 Supplier Thermo Scientific Production Chemicals and Services Supplier No. PZP111WFI036DM55
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 64-17-5 (Ethyl Alcohol)
Molecular Weight (g/mol) 46.07 (Ethyl Alcohol)

Specifications

Quantity 55 Gallon
Packaging Poly Drum
Boiling Point > 188.6 - < 194 °F (> 87 - < 90 °C)
Percent Purity 18% (36 Proof)
Chemical Name or Material Ethyl Alcohol 18% (36 Proof)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.