Learn More
Ethyl Alcohol 40 Proof with WFI, Greenfield
Specifications
Specifications
| CAS | 64-17-5 |
| Quantity | 5 Gallon |
| Packaging | Poly Pail |
| Boiling Point | > 188.6 - < 194 °F (> 87 - < 90 °C) |
| Molecular Weight (g/mol) | 46.07 (Ethanol), 18.02 (Water) |
| Percent Purity | 70% (40 Proof) |
| Grade | ACS/NF/USP |
| Chemical Name or Material | Ethyl Alcohol 20% (40 Proof) |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.