Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl Alcohol 40 Proof with WFI, Greenfield

Catalog No. 502946012
Encompass
Change view
Click to view available options
Quantity:
270 Gallon
5 Gallon
55 Gallon
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-294-6012 270 Gallon
50-294-6096 55 Gallon
50-294-5970 5 Gallon
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 50-294-6012 Supplier Thermo Scientific Production Chemicals and Services Supplier No. P111000040IBC270
Add to Cart
Add to Cart
Request a Quote

Specifications

CAS 64-17-5 (Ethyl Alcohol)
Quantity 270 Gallon
Packaging Tote
Boiling Point > 188.6 - < 194 °F (> 87 - < 90 °C)
Molecular Weight (g/mol) 46.07 (Ethanol), 18.02 (Water)
Percent Purity 20% (40 Proof)
Grade ACS/NF/USP
Chemical Name or Material Ethyl Alcohol 20% (40 Proof)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.