Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl Alcohol, 95% Denatured, ACS Grade, Innovating Science™
SCI_ED

Catalog No. S031125 Shop All Aldon Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S031125 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
WARNING! : This product can expose you to Methanol, which is known to the State of California to cause birth defects or other reproductive harm.
Catalog No. S031125 Supplier Aldon Corporation Supplier No. IS14026
Add to Cart
Add to Cart
WARNING! : This product can expose you to Methanol, which is known to the State of California to cause birth defects or other reproductive harm.

Chemical Identifiers

CAS 64-17-5

Specifications

Quantity 500 mL
Packaging HDPE
Boiling Point 74 to 80°C
Percent Purity 95%
Grade ACS
Chemical Name or Material Ethanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.