Learn More
Electron Microscopy Sciences Ethyl Alcohol, Denatured 1 QT
SDP
Supplier:  Electron Microscopy Sciences 15050
Encompass_Preferred
C₂H₅OH F.W. 46.07
Blended 95% Ethanol and 5% Methanol Suitable for all routine histology/cytology preparations.
Catalog No. 50-980-458