Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl Alcohol Denatured (Proprietary Solvent), Certified, Fisher Chemical™
GSA_VA

Catalog No. A407P4 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
1 L
4 L
20 L
200 L
Packaging:
Glass Bottle
Lined Steel Drum
Poly Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
A407P-4 4 L Poly Bottle
A407-20 20 L Lined Steel Drum
A407-200 200 L Lined Steel Drum
A407-1 1 L Glass Bottle
A407-4 4 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. A407P-4
  • $375.50
    $300.65 / Each

    Save $74.85

    20% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
  • $1,152.00
    $922.65 / Case of 4 EA

    Save $229.35

    20% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart
Edge

missing translation for 'validMainframeMsg'

Formulation: Ethyl Alcohol (100 parts), methyl alcohol (at least 4 parts), methyl iso-butyl ketone (at least 1 part), ethyl acetate (at least 0.87 parts), and rubber hydrocarbon solvent (at least 1 part)

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64-17-5
Molecular Formula C2H6O
Molecular Weight (g/mol) 46.069
MDL Number MFCD00003568
InChI Key LFQSCWFLJHTTHZ-UHFFFAOYSA-N
Synonym ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alcohol, anhydrol, tecsol
PubChem CID 702
ChEBI CHEBI:16236
IUPAC Name ethanol
SMILES CCO

Specifications

Melting Point -90.0°C
Color Colorless
Density 0.7905g/cm³
Quantity 4 L
Flash Point 13.9°C (57°F)
Identification Pass Test
Packaging Poly Bottle
Physical Form Liquid
Specific Gravity 0.7905
Vapor Pressure 40.9mmHg at 20°C
UN Number 1170
Solubility Information Soluble in water
Boiling Point 78.5°C
Grade Certified
Chemical Name or Material Ethyl Alcohol Denatured, Proprietary Solvent
Show More Show Less
Safety handling Solvent Safety handling Check Alert storage code Red
Hazard Statement DANGER!
Precautionary Statement Emergency Overview
Flammable liquid and vapor. Harmful by inhalation, in contact with skin and if swallowed. Exposure through inhalation may result in delayed pulmonary edema, which may be fatal. Aspiration hazard if swallowed - can enter lungs and cause damage. Possible risks of irreversible effects. Causes respiratory tract burns. Irritating to eyes and skin. May cause central nervous system effects. This substance has caused adverse reproductive and fetal effects in humans. Substances known to cause developmental toxicity in humans. WARNING! This product contains a chemical known in the State of California to cause birth defects or. Toxic to aquatic organisms, may cause long-term adverse effects in the aquatic environment. Toxic to aquatic organisms, may cause long-term adverse effects in the aquatic environment. Oxidizing agent.

NFPA

Health:3
Flammability:3
Instability:0
DOTInformation DOT Class 3, : Flammable Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.