Learn More
Sigma Aldrich Fine Chemicals Biosciences Ethyl alcohol denatured re200L
Catalog No. 502905936
Encompass
The product is denatured ethyl alcohol and it contains following as denaturant1% ethyl acetate 1% hydrocarbons 1% methyl isobutyl ketone 5% methanolEthanol a primary alcohol is an important raw material for the synthesis of various compounds. It can be synthesized either from molasses by fermentation or by hydration of ethylene. It acts as a hydrogen source and reaction medium during the transfer hydrogenation of carbonyl compounds in the presence of sodium hydroxide. The impact of ethanol/water fumigation on the performance of diesel engines has been studied. Denatured alcohol is also referred to as industrial methylated spirit.
Catalog No. 50-290-5936 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 187380200L
The product is denatured ethyl alcohol and it contains following as denaturant1% ethyl acetate 1% hydrocarbons 1% methyl isobutyl ketone 5% methanolEthanol a primary alcohol is an important raw material for the synthesis of various compounds. It can be synthesized either from molasses by fermentation or by hydration of ethylene. It acts as a hydrogen source and reaction medium during the transfer hydrogenation of carbonyl compounds in the presence of sodium hydroxide. The impact of ethanol/water fumigation on the performance of diesel engines has been studied. Denatured alcohol is also referred to as industrial methylated spirit.