Learn More
Sigma Aldrich Fine Chemicals Biosciences Ethyl alcohol, Pure, 64-17-5, MFCD00003568, 4L
Catalog No. 501657088
Encompass
Linear Formula CH3CH2OH, ≥99.5%, Molecular Weight 46.07, 200 proof, ACS reagent, Synonym: Ethanol, absolute alcohol, non-denatured ethanol
This item is not returnable. View return policy
Catalog No. 50-165-7088 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 4598444L
Linear Formula CH3CH2OH, ≥99.5%, Molecular Weight 46.07, 200 proof, ACS reagent, Synonym: Ethanol, absolute alcohol, non-denatured ethanol